siRNA Details
Gene Symbol: LOC102723838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135076.1 | 1, 2 | 112 | |||
GenBank mRNA | BX093783.1 | - | 1, 2 | 112 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000529278.1 | - | 1, 2 | 112 | Chr. 11 - 102467255 - 102498801 [+] on Build GRCh38 |
