siRNA Details
| Gene Symbol: LINC00681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102423.1 | 2 | 424 | |||
| GenBank mRNA | BM725196.1 | - | 2 | 424 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000530150.1 | - | 2 | 424 | Chr. 8 - 12794243 - 12818291 [+] on Build GRCh38 | |

LINC00681
Genomic Map