siRNA Details
| Gene Symbol: LOC100507144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120528.1 | 3 | 989 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000528366.1 | - | 3 | 989 | Chr. 11 - 35132655 - 35138032 [-] on Build GRCh38 | |

LOC100507144
Genomic Map