siRNA Details
| Gene Symbol: LOC283177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033852.1 | 2 | 1217 | |||
| GenBank mRNA | AK095081.1 | - | 2 | 1101 | ||
| BC112333.1 | - | 2 | 964 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283177| Gene Symbol: LOC283177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033852.1 | 2 | 1217 | |||
| GenBank mRNA | AK095081.1 | - | 2 | 1101 | ||
| BC112333.1 | - | 2 | 964 | |||