siRNA Details
| Gene Symbol: FAM222A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026661.2 | 3, 4 | 485 | |||
| NR_026662.2 | 2, 3 | 424 | ||||
| GenBank mRNA | BC007821.1 | - | 2, 3 | 425 | ||
| BQ684893.1 | - | 2, 3 | 446 | |||
| DA082338.1 | - | 3, 4 | 482 | |||

FAM222A-AS1
Genomic Map