siRNA Details
| Gene Symbol: JRKL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047481.1 | 4 | 464 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000511243.2 | - | 4 | 414 | Chr. 11 - 96447132 - 96506826 [-] on Build GRCh38 | |

JRKL-AS1
Genomic Map