siRNA Details
| Gene Symbol: JRKL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047481.1 | 3, 4 | 370 | |||
| GenBank mRNA | BE972282.1 | - | 3, 4 | 371 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000511243.2 | - | 3, 4 | 320 | Chr. 11 - 96447132 - 96506826 [-] on Build GRCh38 | |

JRKL-AS1
Genomic Map