siRNA Details
| Gene Symbol: IFNG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104124.1 | 5 | 1845 | |||
| NR_104125.1 | 4 | 1727 | ||||
| GenBank mRNA | AK124066.1 | - | 4 | 1727 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000538665.5 | - | 4 | 1727 | Chr. 12 - 67989445 - 68021327 [+] on Build GRCh38 | |

IFNG-AS1
Genomic Map