siRNA Details
| Gene Symbol: IFNG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104124.1 | 5 | 673 | |||
| NR_104125.1 | 4 | 555 | ||||
| GenBank mRNA | AK124066.1 | - | 4 | 555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFNG-AS1| Gene Symbol: IFNG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104124.1 | 5 | 673 | |||
| NR_104125.1 | 4 | 555 | ||||
| GenBank mRNA | AK124066.1 | - | 4 | 555 | ||