siRNA Details
Gene Symbol: LINC01479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120456.1 | 2 | 231 | |||
GenBank mRNA | BM850770.1 | - | 2 | 231 | ||
BX107064.1 | - | 2 | 203 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120456.1 | 2 | 231 | |||
GenBank mRNA | BM850770.1 | - | 2 | 231 | ||
BX107064.1 | - | 2 | 203 |