siRNA Details
| Gene Symbol: LOC105369595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134624.1 | 1 | 152 | |||
| GenBank mRNA | T68370.1 | - | 1 | 152 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000538943.1 | - | 1 | 152 | Chr. 12 - 585865 - 586486 [-] on Build GRCh38 | |

LOC105369595
Genomic Map