siRNA Details
| Gene Symbol: LOC101927637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120451.1 | 4 | 504 | |||
| GenBank mRNA | BU567943.1 | - | 3 | 454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927637| Gene Symbol: LOC101927637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120451.1 | 4 | 504 | |||
| GenBank mRNA | BU567943.1 | - | 3 | 454 | ||