siRNA Details
| Gene Symbol: LINC01152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110124.1 | 3 | 935 | |||
| NR_110132.1 | 3 | 425 | ||||
| GenBank mRNA | D43770.1 | - | LIT1584 CMPD |
3 | 773 | |
| DA197482.1 | - | 3 | 425 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000472655.3 | - | 935 | Chr. 17 - 72030654 - 72038265 [+] on Build GRCh38 | ||

LINC01152
Genomic Map