siRNA Details
Gene Symbol: LINC00507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046392.1 | 5 | 1926 | |||
GenBank mRNA | BC037880.2 | - | 4 | 1728 | ||
BC045798.1 | - | 4 | 1728 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046392.1 | 5 | 1926 | |||
GenBank mRNA | BC037880.2 | - | 4 | 1728 | ||
BC045798.1 | - | 4 | 1728 |