siRNA Details
| Gene Symbol: LOC105369507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135102.1 | 2 | 511 | |||
| NR_135103.1 | 2 | 514 | ||||
| GenBank mRNA | BX106383.1 | - | 2 | 511 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000543182.1 | - | 2 | 511 | Chr. 11 - 115396756 - 115397658 [+] on Build GRCh38 | |

LOC105369507
Genomic Map