siRNA Details
| Gene Symbol: FLJ37505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033987.1 | 3 | 1073 | |||
| GenBank mRNA | AK094824.1 | - | 3 | 1042 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000614177.1 | - | 1046 | Chr. - - [-] on Build | ||

FLJ37505
Genomic Map