siRNA Details
| Gene Symbol: LOC105369691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135030.1 | 1 | 168 | |||
| NR_135031.1 | 1 | 168 | ||||
| GenBank mRNA | HY231162.1 | - | 1 | 207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369691| Gene Symbol: LOC105369691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135030.1 | 1 | 168 | |||
| NR_135031.1 | 1 | 168 | ||||
| GenBank mRNA | HY231162.1 | - | 1 | 207 | ||