siRNA Details
| Gene Symbol: LOC105370424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135255.1 | 1 | 37 | |||
| GenBank mRNA | BC034423.1 | - | 1 | 34 | ||
| BI917327.1 | - | 1 | 37 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000535153.2 | - | 1 | 37 | Chr. 14 - 28783491 - 28785787 [-] on Build GRCh38 | |

LOC105370424
Genomic Map