siRNA Details
| Gene Symbol: KCNK4-TEX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133662.1 | 7 | 1770 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000539086.5 | - | 7 | 1770 | Chr. 11 - 64291722 - 64304769 [+] on Build GRCh38 | |

KCNK4-TEX40
Genomic Map