siRNA Details
Gene Symbol: LOC102724421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120490.1 | 1 | 156 | |||
GenBank mRNA | BX100562.1 | - | 1 | 158 | ||
DB097934.1 | - | 1 | 278 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120490.1 | 1 | 156 | |||
GenBank mRNA | BX100562.1 | - | 1 | 158 | ||
DB097934.1 | - | 1 | 278 |