siRNA Details
Gene Symbol: LOC105369635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135032.1 | 2 | 154 | |||
GenBank mRNA | BX094730.1 | - | 2 | 153 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000537269.1 | - | 2 | 153 | Chr. 12 - 6963246 - 6964447 [+] on Build GRCh38 |
