siRNA Details
Gene Symbol: LOC105369739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135028.1 | 1, 2 | 64 | |||
GenBank mRNA | BX096171.1 | - | 1, 2 | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000550177.2 | - | 1, 2 | 64 | Chr. 12 - 42692216 - 42716851 [+] on Build GRCh38 |
