siRNA Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042722.1 | - | 2 | 1992 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000547824.1 | - | 2 | 1992 | Chr. 12 - 42459366 - 42466128 [+] on Build GRCh38 | |

PRICKLE1
Genomic Map