siRNA Details
| Gene Symbol: LOC101927653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120452.1 | 6 | 1237 | |||
| GenBank mRNA | AK093124.1 | - | 6 | 1237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000550678.1 | - | 6 | 1237 | Chr. 12 - 58565960 - 58781716 [-] on Build GRCh38 | |

LOC101927653
Genomic Map