siRNA Details
Gene Symbol: LOC101927653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120452.1 | 3 | 496 | |||
GenBank mRNA | AK093124.1 | - | 3 | 496 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120452.1 | 3 | 496 | |||
GenBank mRNA | AK093124.1 | - | 3 | 496 |