siRNA Details
Gene Symbol: LOC100506551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103809.1 | 4 | 3210 | |||
GenBank mRNA | AK127077.1 | - | 4 | 3210 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000548738.1 | - | 4 | 3210 | Chr. 12 - 116977442 - 116987337 [-] on Build GRCh38 |
