siRNA Details
| Gene Symbol: LINC01475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120618.1 | 3 | 386 | |||
| GenBank mRNA | DQ372722.1 | - | 3 | 390 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000548010.1 | - | 3 | 386 | Chr. 10 - 99526350 - 99531177 [-] on Build GRCh38 | |

LINC01475
Genomic Map