siRNA Details
Gene Symbol: LINC01475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120618.1 | 4 | 618 | |||
GenBank mRNA | DQ372722.1 | - | 4 | 622 | ||
DQ372723.1 | - | 2 | 189 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120618.1 | 4 | 618 | |||
GenBank mRNA | DQ372722.1 | - | 4 | 622 | ||
DQ372723.1 | - | 2 | 189 |