siRNA Details
| Gene Symbol: MIR4307HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110041.1 | 2 | 275 | |||
| GenBank mRNA | BX115853.1 | - | 2 | 275 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000552926.1 | - | 2 | 245 | Chr. 14 - 26873163 - 26873652 [+] on Build GRCh38 | |

MIR4307HG
Genomic Map