siRNA Details
Gene Symbol: MIR4307HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110041.1 | 1 | 150 | |||
GenBank mRNA | BX115853.1 | - | 1 | 150 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000552926.1 | - | 1 | 120 | Chr. 14 - 26873163 - 26873652 [+] on Build GRCh38 |
