siRNA Details
| Gene Symbol: LOC105369891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135019.1 | 3 | 370 | |||
| GenBank mRNA | BX090745.1 | - | 3 | 370 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000547094.1 | - | 3 | 370 | Chr. 12 - 89947693 - 89949726 [+] on Build GRCh38 | |

LOC105369891
Genomic Map