siRNA Details
| Gene Symbol: LINC01490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120468.1 | 5 | 1238 | |||
| NR_120469.1 | 5 | 1182 | ||||
| NR_120470.1 | 3 | 1034 | ||||
| GenBank mRNA | BC037383.1 | - | 3 | 1038 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000551399.1 | - | 3 | 1034 | Chr. 12 - 80763154 - 80770717 [+] on Build GRCh38 | |

LINC01490
Genomic Map