siRNA Details
Gene Symbol: LINC01490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120468.1 | 5 | 1158 | |||
NR_120469.1 | 5 | 1102 | ||||
NR_120470.1 | 3 | 954 | ||||
GenBank mRNA | BC037383.1 | - | 3 | 958 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000551399.1 | - | 3 | 954 | Chr. 12 - 80763154 - 80770717 [+] on Build GRCh38 |
