siRNA Details
Gene Symbol: LOC101928449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110089.1 | 1 | 395 | |||
GenBank mRNA | BC031963.1 | - | 1 | 395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000550506.1 | - | 1 | 395 | Chr. 12 - 81953719 - 81993133 [+] on Build GRCh38 |
