siRNA Details
| Gene Symbol: LOC101929084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110103.1 | 2 | 188 | |||
| GenBank mRNA | BC041342.2 | - | 2 | 188 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000550185.5 | - | 2 | 188 | Chr. 12 - 103547794 - 103559815 [+] on Build GRCh38 | |

LOC101929084
Genomic Map