siRNA Details
| Gene Symbol: LOC101927292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110048.1 | 6 | 2998 | |||
| GenBank mRNA | BC034605.1 | - | 6 | 2998 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000546821.5 | - | 6 | 2998 | Chr. 12 - 49911953 - 49926339 [+] on Build GRCh38 | |

LOC101927292
Genomic Map