siRNA Details
Gene Symbol: LINC01481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120460.1 | 4 | 239 | |||
NR_120461.1 | 4 | 356 | ||||
GenBank mRNA | DB462634.1 | - | 4 | 345 | ||
HY002672.1 | - | 4 | 488 | |||
HY013176.1 | - | 4 | 239 | |||
HY048577.1 | - | 4 | 350 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000550216.1 | - | 4 | 345 | Chr. 12 - 70224993 - 70243349 [-] on Build GRCh38 |
