siRNA Details
| Gene Symbol: LINC01481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120461.1 | 4, 5 | 473 | |||
| GenBank mRNA | DB462634.1 | - | 4, 5 | 462 | ||
| HY048577.1 | - | 4, 5 | 467 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000550216.1 | - | 4, 5 | 462 | Chr. 12 - 70224993 - 70243349 [-] on Build GRCh38 | |

LINC01481
Genomic Map