siRNA Details
Gene Symbol: LOC414300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121211.1 | 2 | 1090 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000550088.1 | - | 2 | 627 | Chr. 12 - 105102472 - 105107179 [-] on Build GRCh38 |
