siRNA Details
| Gene Symbol: LOC101928137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110130.1 | 4 | 739 | |||
| GenBank mRNA | AK310448.1 | - | 4 | 741 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000550723.1 | - | 4 | 739 | Chr. 12 - 73159190 - 73208317 [+] on Build GRCh38 | |

LOC101928137
Genomic Map