siRNA Details
Gene Symbol: LOC283332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026948.1 | 3 | 872 | |||
GenBank mRNA | AK093215.1 | - | 3 | 866 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000550636.1 | - | 3 | 866 | Chr. 12 - 49908893 - 49911857 [-] on Build GRCh38 |
