siRNA Details
| Gene Symbol: LOC101929162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120474.1 | 4, 5 | 1521 | |||
| GenBank mRNA | AK093309.1 | - | 4, 5 | 1487 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000546714.1 | - | 4, 5 | 1487 | Chr. 12 - 107736589 - 107759968 [+] on Build GRCh38 | |

LOC101929162
Genomic Map