siRNA Details
Gene Symbol: LINC00485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033855.1 | 5 | 2661 | |||
GenBank mRNA | BC037211.1 | - | 5 | 2660 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000547179.1 | - | 5 | 2661 | Chr. 12 - 102809280 - 102824399 [-] on Build GRCh38 |
