siRNA Details
| Gene Symbol: LOC105369860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135021.1 | 1 | 125 | |||
| GenBank mRNA | BC034620.1 | - | 1 | 97 | ||
| BC045810.1 | - | 1 | 125 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369860| Gene Symbol: LOC105369860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135021.1 | 1 | 125 | |||
| GenBank mRNA | BC034620.1 | - | 1 | 97 | ||
| BC045810.1 | - | 1 | 125 | |||