siRNA Details
| Gene Symbol: LOC105369980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135001.1 | 1 | 210 | |||
| NR_135002.1 | 1 | 210 | ||||
| GenBank mRNA | DA146183.1 | - | 1 | 149 | ||
| DA148561.1 | - | 1 | 204 | |||
| DA302221.1 | - | 1 | 203 | |||
| HY021340.1 | - | 1 | 210 | |||

LOC105369980
Genomic Map