siRNA Details
Gene Symbol: LINC00592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027358.1 | 5 | 934 | |||
GenBank mRNA | BC039104.1 | - | 5 | 934 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000549830.2 | - | 5 | 934 | Chr. 12 - 52210930 - 52223804 [+] on Build GRCh38 |
