siRNA Details
| Gene Symbol: TESC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120464.1 | 2 | 327 | |||
| GenBank mRNA | HY007462.1 | - | 2 | 327 | ||
| HY052120.1 | - | 2 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TESC-AS1| Gene Symbol: TESC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120464.1 | 2 | 327 | |||
| GenBank mRNA | HY007462.1 | - | 2 | 327 | ||
| HY052120.1 | - | 2 | 289 | |||