siRNA Details
Gene Symbol: LOC105369920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135016.1 | 1 | 157 | |||
GenBank mRNA | AI792597.1 | - | 1 | 157 | ||
BX108644.1 | - | 1 | 157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553095.1 | - | 1 | 157 | Chr. 12 - 95803097 - 95823307 [+] on Build GRCh38 |
