siRNA Details
| Gene Symbol: LOC100506603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104183.1 | 3 | 859 | |||
| NR_104184.1 | 2 | 700 | ||||
| GenBank mRNA | AK125727.1 | - | 2 | 699 | ||
| BI907467.1 | - | 3 | 287 | |||
| DA369488.1 | - | 2 | 495 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506603| Gene Symbol: LOC100506603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104183.1 | 3 | 859 | |||
| NR_104184.1 | 2 | 700 | ||||
| GenBank mRNA | AK125727.1 | - | 2 | 699 | ||
| BI907467.1 | - | 3 | 287 | |||
| DA369488.1 | - | 2 | 495 | |||