siRNA Details
Gene Symbol: LOC101927267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120449.1 | 8 | 4079 | |||
GenBank mRNA | AK128056.1 | - | 8 | 4079 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553259.1 | - | 8 | 4079 | Chr. 12 - 49292631 - 49324576 [-] on Build GRCh38 |
